missing translation for 'onlineSavingsMsg'
Learn More

PAGE3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 24598-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16893085

  • 349.00€ / 150µL
Expédition estimée: 16-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

PAGE-3 (prostate-associated gene 3 protein), also known as GAGED1 (putative G antigen family Dmember 1), is a 113 amino acid protein that belongsto the GAGE family. The gene that encodes PAGE-3 consists of approximately 6,432 bases andmapsto human chromosome Xp11.21. Chromosome X consists of about 153million base pairs and nearly 1,000 genes. The combination of a X and Y chromosome lead to normal male development while two copies of X lead to normal female development. There are a number ofconditionsrelated to an unusual number and combination ofsexchromosomes being inherited, including Turner′s syndrome, Klinefelter′ssyndrome and Triple X syndrome. Color blindness, hemophilia, and Duchennemuscular dystrophy arewellknown X chromosome-linked conditionswhich affectmalesmore frequently as males carry a single X chromosome.
Spécification

Spécification

PAGE3
Polyclonal
Unconjugated
PAGE3
GAGEC1, GAGED1, PAGE 3, PAGE3
Rabbit
Antigen Affinity Chromatography
RUO
139793
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5JUK9
PAGE3
PAGE3 Fusion Protein Ag20171
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions