missing translation for 'onlineSavingsMsg'
Learn More

PARP1 Mouse anti-Human, Mouse, Rat, Clone: 1D7D4, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66520-1-IG-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16828475

  • 121.38€ / 20µL
Expédition estimée: 27-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody was generated against the N-terminal region of human PARP1 and it recognizes the full-length as well as the cleavage of the PARP1.

Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.
Spécification

Spécification

PARP1
Monoclonal
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P09874, P11103, P27008
PARP1
PARP1 Fusion Protein Ag19173
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG1
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot
1D7D4
Unconjugated
PARP1
ADPRT, ADPRT 1, ADPRT1, pADPRT 1, PARP, PARP 1, PARP1, poly (ADP ribose) polymerase 1, Poly [ADP ribose] polymerase 1, Poly[ADP ribose] synthase 1, PPOL
Mouse
Protein G
RUO
11545, 142, 25591
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions