missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCAF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13983-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Mixed-lineage kinases (MLKs) function as mitogen-activated protein kinase kinase kinases (MAP3Ks), activating the p38 and JNK signaling cascades. Systematic mutational analyses of tyrosine kinases reveal that a minimum of 30% of colorectal cancer contain at least one mutation in the tyrosine kinases; sampling of colorect cancer cells suggest an important role of MLK4 in this process.Spécification
PCAF | |
Polyclonal | |
Unconjugated | |
KAT2B | |
CAF, Histone acetylase PCAF, Histone acetyltransferase PCAF, K(lysine) acetyltransferase 2B, KAT2B, Lysine acetyltransferase 2B, P, P/CAF, P300/CBP associated factor, PCAF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18519, 301164, 8850 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92831, Q9JHD1 | |
Kat2b | |
PCAF Fusion Protein Ag5028 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |