missing translation for 'onlineSavingsMsg'
Learn More
Learn More
persephin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 19713-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The antibody is specific to PSPN.
Exhibits neurotrophic activity on mesencephalic dopaminergic and motor neurons.Spécification
persephin | |
Polyclonal | |
Unconjugated | |
PSPN | |
persephin, PSP, PSPN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5623 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60542 | |
PSPN | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |