missing translation for 'onlineSavingsMsg'
Learn More

PIAS1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 23395-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16832335

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

PIAS1 functions as an E3-type small ubiquitin-like modifier (SUMO) ligase, stabilizing the interaction between UBE2I and the substrate, and as a SUMO-tethering factor. This protein plays a crucial role in transcriptional coregulation of various cellular pathways, including the STAT pathway, the p53 pathway and the steroid hormone signaling pathway. It functions in testis as a nuclear receptor transcriptional coregulator and may have a role in androgen receptor initiation and maintece of spermatogenesis. The effects of transcriptional coregulation, transactivation or silencing, may vary depending upon the biological context.
Spécification

Spécification

PIAS1
Polyclonal
Unconjugated
PIAS1
DDXBP1, DEAD/H box binding protein 1, E3 SUMO protein ligase PIAS1, Gu binding protein, GU/RH II, PIAS1, ZMIZ3
Rabbit
Antigen Affinity Chromatography
RUO
8554
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75925
PIAS1
PIAS1 Fusion Protein Ag18132
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions