missing translation for 'onlineSavingsMsg'
Learn More

PKLR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 22456-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831945

  • 121.38€ / 20µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a pyruvate kinase that catalyzes the transphosphorylation of phohsphoenolpyruvate into pyruvate and ATP, which is the rate-limiting step of glycolysis. Defects in this enzyme, due to gene mutations or genetic variations, are the common cause of chronic hereditary nonspherocytic hemolytic anemia (CNSHA or HNSHA). Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

PKLR
Polyclonal
Unconjugated
PKLR
PK1, PKL, PKLR, PKR, PKRL, Pyruvate kinase 1, Pyruvate kinase isozymes R/L, pyruvate kinase, liver and RBC, R type/L type pyruvate kinase, Red cell/liver pyruvate kinase, RPK
Rabbit
Antigen Affinity Chromatography
RUO
18770, 24651, 5313
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.61 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P12928, P30613, P53657
Pklr
PKLR Fusion Protein Ag17933
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions