missing translation for 'onlineSavingsMsg'
Learn More

PPM1D Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26532-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16834965

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a type IV membrane protein. It is present in the plasma membrane and intracellular vesicles. It may also be associated with the cytoskeleton. This protein may function in vesicle trafficking, membrane fusion, protein complex assembly and cell motility. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Spécification

Spécification

PPM1D
Polyclonal
Unconjugated
PPM1D
PP2C DELTA, PPM1D, Protein phosphatase 1D, WIP1
Rabbit
Antigen Affinity Chromatography
RUO
53892, 8493
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15297, Q9QZ67
Ppm1d
PPM1D Fusion Protein Ag24751
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions