missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 18203-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is specific to PPM1L.
PPM1L, or PP2CE, belongs to the PP2C group of serine/threonine phosphatases, which are distinguished from other phosphatases by their structure, absolute requirement for Mg(2+) or Mn(2+), and insensitivity to okadaic acid. PP2Cs regulate stress-activated protein kinase (SAPK.Spécification
PPM1L | |
Polyclonal | |
Unconjugated | |
PPM1L | |
PP2C epsilon, PP2CE, PPM1 LIKE, PPM1L, Protein phosphatase 1 like, Protein phosphatase 1L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
151742, 242083, 310506 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5SGD2, Q8BHN0 | |
Ppm1l | |
PPM1L Fusion Protein Ag12775 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |