Learn More
PREPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 12478-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16830214
  • 140.00€ / 20µL
Expédition estimée: 29-04-2024
Description
The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
Spécification
Spécification
PREPL
Polyclonal
Unconjugated
PREPL
KIAA0436, PPCEL, PREPL, prolyl endopeptidase like, Prolylendopeptidase like
Rabbit
Antigen Affinity Chromatography
RUO
213760, 298771, 9581
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q4J6C6, Q5HZA6, Q8C167
PREPL
PREPL Fusion Protein Ag3196
20 μL
Primary
Mouse, Human, Rat