Learn More
PRKDC Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28534-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The PRKDC antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PRKDC. This antibody recognizes human antigen. The PRKDC antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
PRKDC | |
Polyclonal | |
Unconjugated | |
150 μL | |
5591 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
200 μg/mL | |
Rabbit | |
Primary | |
Human |