missing translation for 'onlineSavingsMsg'
Learn More

PRPF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10728-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16827303

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF4 is 1 of several proteins that associate with U4 and U6 snRNPs.
Spécification

Spécification

PRPF4
Polyclonal
Unconjugated
PRPF4
HPRP4, HPRP4P, PRP4, PRP4 homolog, Prp4p, PRPF4, U4/U6 snRNP 60 kDa protein, WD splicing factor Prp4
Rabbit
Antigen Affinity Chromatography
RUO
298095, 70052, 9128
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43172, Q9DAW6
Prpf4
PRPF4 Fusion Protein Ag1193
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions