Learn More
PRTN3 Mouse anti-Human, Unconjugated, Clone: 2D1B8, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67030-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The PRTN3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRTN3. This antibody recognizes human antigen. The PRTN3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
PRTN3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2D1B8 | |
Unconjugated | |
20 μL | |
5657 | |
IgG2b |