Learn More
PRTN3 Mouse anti-Human, Unconjugated, Clone: 2D1B8, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67030-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16829415
  • 140.00€ / 20µL
Expédition estimée: 06-05-2024
Description
The PRTN3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRTN3. This antibody recognizes human antigen. The PRTN3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
Spécification
PRTN3
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
2D1B8
Unconjugated
20 μL
5657
IgG2b