missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTEN (Tumor Suppressor Oncoprotein) Mouse, Clone: PTEN/2110, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 5278-MSM1-P0
Informations supplémentaires : Poids : 0.01000kg
Spécification
PTEN (Tumor Suppressor Oncoprotein) | |
Cytoplasmic and Nuclear | |
PTEN/2110 | |
Unconjugated | |
Mouse | |
55kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
5278 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human PTEN protein | |
MCF-7 or HeLa Cells. Prostate, Breast, Liver, Renal Carcinoma. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |