missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPMT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11493-1-AP-150UL
Additional Details : Poids : 0.00010kg
Description
Lipid phosphatase which dephosphorylates phosphatidylglycerophosphate (PGP) to phosphatidylglycerol (PG). PGP is an essential intermediate in the biosynthetic pathway of cardiolipin, a mitochondrial-specific phospholipid regulating the membrane integrity and activities of the organelle. Has also been shown to display phosphatase activity toward phosphoprotein substrates, specifically mediates dephosphorylation of mitochondrial proteins, thereby playing an essential role in ATP production. Has probably a preference for proteins phosphorylated on Ser and/or Thr residues compared to proteins phosphorylated on Tyr residues. Probably involved in regulation of insulin secretion in pancreatic beta cells.Specifications
PTPMT1 | |
Polyclonal | |
Unconjugated | |
PTPMT1 | |
DUSP23, MOSP, PLIP, PNAS 129, PTEN like phosphatase, PTPMT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114971, 29390, 66461 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WUK0 | |
Ptpmt1 | |
PTPMT1 Fusion Protein Ag2063 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |