missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRZ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55125-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is specific to PTPRZ1.
Protein tyrosine phosphatase that negatively regulates oligodendrocyte precursor proliferation in the embryonic spinal cord. Required for normal differentiation of the precursor cells into mature, fully myelinating oligodendrocytes. May play a role in protecting oligondendrocytes against apoptosis. May play a role in the establishment of contextual memory, probably via the dephosphorylation of proteins that are part of important signaling cascades. .Spécification
PTPRZ1 | |
Polyclonal | |
Unconjugated | |
PTPRZ1 | |
HPTPZ, HPTPzeta, HTPZP2, phosphacan, PTP ZETA, PTP18, PTPRZ, PTPRZ1, PTPRZ2, PTPZ, R PTP zeta, R PTP zeta 2, RPTPB, RPTPbeta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19283, 25613, 5803 | |
-20°C | |
Liquid |
Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B9EKR1, P23471, Q62656 | |
PTPRZ1 | |
Peptide | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |