missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10735-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
RAC, a small GTPase in the ras family, regulates the formation of membrane ruffles, lamellipodia and filopodia. It is involved in cytoskeletal actin organization and transformation. The p21 activated kinases (PAKs) are direct targets of active Rac and Cdc42 which can induce the assembly of polarized cytoskeletal structures when expressed in fibroblasts. RAC1 and RAC2 are also essential components of NADPH oxidase, the enzyme responsible for generating free radicals.Spécification
RAC2 | |
Polyclonal | |
Unconjugated | |
RAC2 | |
EN 7, Gx, HSPC022, p21 Rac2, RAC2, Small G protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5880 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P15153 | |
RAC2 | |
RAC2 Fusion Protein Ag1202 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |