missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27071-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.Spécification
RAD21 | |
Polyclonal | |
Unconjugated | |
RAD21 | |
hHR21, HR21, HRAD21, KIAA0078, MCD1, Nuclear matrix protein 1, NXP 1, NXP1, RAD21, RAD21 homolog (S. pombe), SCC1, SCC1 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5885 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60216 | |
RAD21 | |
RAD21 Fusion Protein Ag25481 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |