missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17988-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
REEP1 belongs to the DP1 family and may enhance the cell surface expression of odorant receptors.Spécification
REEP1 | |
Polyclonal | |
Unconjugated | |
REEP1 | |
C2orf23, receptor accessory protein 1, REEP1, SPG31 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362384, 52250, 65055 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BGH4, Q9H902 | |
Reep1 | |
REEP1 Fusion Protein Ag12418 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |