missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNPC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25820-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
U11/U12 snRNP 65K, also known as RBM40 (RNA-binding protein 40), RNPC3 (RNA-binding region-containing protein 3) or RNP, is a 517 amino acid nuclear protein that plays a role in pre-mRNA U12-dependent splicing by the minor spliceosome, ultimately leading to the removal of U12-type introns. Expressed at highest levels in kidney and pancreas, U11/U12 snRNP 65K is also weakly detected in placenta, heart, brain, lung, liver, thymus, colon, ovary, testis, prostate, spleen and small intestine. The gene encoding U11/U12 snRNP 65K maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson′s disease, Gaucher disease, schizophrenia and Usher syndrome.Spécification
RNPC3 | |
Polyclonal | |
Unconjugated | |
RNPC3 | |
KIAA183, KIAA1839, RBM40, RNA binding motif protein 40, RNA binding protein 40, RNP, RNPC3, U11/U12 65K, U11/U12 snRNP 65 kDa protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55599, 67225 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3UZ01, Q96LT9 | |
RNPC3 | |
RNPC3 Fusion Protein Ag22957 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |