missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SALL4 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24500-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Sall4 is a probable transcription factor. Defects in Sall4 are the cause of Okihiro syndrome; also known as Duane radial ray syndrome (DRRS). It is a disorder characterized by the association of forearm malformations with Duane retraction syndrome. Sall4 is also involved in forelimb and heart development in mice.Spécification
SALL4 | |
Polyclonal | |
Unconjugated | |
SALL4 | |
DRRS, HSAL4, sal like 4 (Drosophila), Sal like protein 4, SALL4, Zinc finger protein SALL4, ZNF797 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57167, 686412 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UJQ4 | |
SALL4 | |
SALL4 Fusion Protein Ag17480 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |