Learn More
SEC24D Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 13673-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16832014
  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 
Description
Description
The SEC24D antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SEC24D. This antibody recognizes human, mouse antigen. The SEC24D antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
Spécification
SEC24D
Polyclonal
Unconjugated
150 μL
9871
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
267 μg/mL
Rabbit
Primary
Human, Mouse