Learn More
Secretogranin V Mouse anti-Human, Unconjugated, Clone: 1B9H6, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67406-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16840016
  • 140.00€ / 20µL
Expédition estimée: 15-05-2024
Description
The Secretogranin V antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Secretogranin V. This antibody recognizes human antigen. The Secretogranin V antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
Spécification
Secretogranin V
Monoclonal
927 μg/mL
Mouse
Primary
Human
Immunohistochemistry, ELISA
1B9H6
Unconjugated
20 μL
6447
IgG1