Learn More
Secretogranin V Mouse anti-Human, Unconjugated, Clone: 1B9H6, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67406-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Secretogranin V antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Secretogranin V. This antibody recognizes human antigen. The Secretogranin V antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
Secretogranin V | |
Monoclonal | |
927 μg/mL | |
Mouse | |
Primary | |
Human |
Immunohistochemistry, ELISA | |
1B9H6 | |
Unconjugated | |
150 μL | |
6447 | |
IgG1 |