Learn More
SERCA2 ATP2A2 Mouse anti-Human, Unconjugated, Clone: 1F11A11, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67248-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16829775
  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 
Description
Description
The SERCA2, ATP2A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SERCA2, ATP2A2. This antibody recognizes human, mouse, rat antigen. The SERCA2, ATP2A2 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
SERCA2 ATP2A2
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
Western Blot
1F11A11
Unconjugated
20 μL
488
IgG1