missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SESN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11431-2-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
May function as an intracellular leucine sensor that negatively regulates the TORC1 signaling pathway. May also regulate the insulin-receptor signaling pathway through activation of TORC2. This metabolic regulator may also play a role in protection against oxidative and genotoxic stresses.Spécification
SESN3 | |
Polyclonal | |
Unconjugated | |
SESN3 | |
SESN3, SEST3, sestrin 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
143686, 315427, 75747 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P58005, Q9CYP7 | |
Sesn3 | |
SESN3 Fusion Protein Ag1961 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |