missing translation for 'onlineSavingsMsg'
Learn More

SETD8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14063-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16832534

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Histone methyltransferase that specifically monomethylates ′Lys-20′ of histone H4. H4 ′Lys-20′ monomethylation is enriched during mitosis and represents a specific tag for epigenetic transcriptional repression. Mainly functions in euchromatin regions, thereby playing a central role in the silencing of euchromatic genes. Required for cell proliferation, probably by contributing to the maintece of proper higher order structure of DNA during mitosis. Involved in chromosome condensation and proper cytokinesis. Nucleosomes are preferred as substrate compared to free histones.
Spécification

Spécification

SETD8
Polyclonal
Unconjugated
KMT5A
H4 K20 HMTase SETD8, KMT5A, Lysine N methyltransferase 5A, PR Set7, PR/SET07, PRSET7, SET07, SET8, SETD8
Rabbit
Antigen Affinity Chromatography
RUO
387893, 67956
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q2YDW7, Q9NQR1
KMT5A
SETD8 Fusion Protein Ag5114
20 μL
Primary
Human, Mouse
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions