missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRP2 Mouse anti-Human, Rat, Clone: 1F3A1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66328-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. Methylation of this gene is a potential marker for the presence of colorectal cancer.Spécification
SFRP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96HF1 | |
SFRP2 | |
SFRP2 Fusion Protein Ag18840 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1F3A1 | |
Unconjugated | |
SFRP2 | |
FRP 2, FRP2, SARP 1, SARP1, SDF 5, sFRP 2, SFRP2, UNQ361/PRO697 | |
Mouse | |
Protein G | |
RUO | |
310552, 6423 | |
-20°C | |
Liquid |