missing translation for 'onlineSavingsMsg'
Learn More

SGK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 12699-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16830564

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene is a member of the Ser/Thr protein kinase family and encodes a phosphoprotein with a PX (phox homology) domain. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Spécification

Spécification

SGK3
Polyclonal
Unconjugated
SGK3
CISK, SGK3, SGKL
Rabbit
Antigen Affinity Chromatography
RUO
170755, 171498, 23678
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8R4V0, Q96BR1, Q9ERE3
Sgk3
SGK3 Fusion Protein Ag3149
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions