missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3GL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27014-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Implicated in endocytosis. May recruit other proteins to membranes with high curvature.Spécification
SH3GL1 | |
Polyclonal | |
Unconjugated | |
SH3GL1 | |
CNSA1, EEN, EEN fusion partner of MLL, Endophilin 2, Endophilin A2, SH3 domain GRB2 like 1, SH3 domain protein 2B, SH3D2B, SH3GL1, SH3P8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20405, 6455, 81922 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35964, Q62419, Q99961 | |
SH3GL1 | |
SH3GL1 Fusion Protein Ag25550 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |