Learn More
SIN3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14638-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16803384
  • 140.00€ / 20µL
Expédition estimée: 30-05-2024
Description
Mammalian Sin3 (mSin3) is a 150 kDa protein closely related to the yeast SIN3 repressor protein involved in the transcriptional repression of many genes. There are two mammalian isoforms, mSin3A and mSin3B. Containing 4 paired amphipathic helix domains (PAH domains), mSin3A and mSin3B have been shown to directly interact with several other transcriptional repressor proteins including HDAC1, HDAC2, RbAp46, the methyl CpG binding protein MeCP2, the Mad/Max heterodimer, and the corepressors silencing mediator of retinoic acid & thyroid hormone receptor (SMRT) and nuclear receptor corepressor (N-CoR).
Spécification
Spécification
SIN3A
Polyclonal
Unconjugated
SIN3A
KIAA0700, SIN3A
Rabbit
Antigen Affinity Chromatography
RUO
20466, 25942, 363067
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q60520, Q96ST3
Sin3a
SIN3A Fusion Protein Ag6219
20 μL
Primary
Rat, Human, Mouse