Learn More
SIRT5 Mouse anti-Human, Unconjugated, Clone: 1F2B2, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67257-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16829785
  • 121.38€ / 20µL
Expédition estimée: 06-10-2022
Description
The SIRT5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SIRT5. This antibody recognizes human antigen. The SIRT5 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
SIRT5
Monoclonal
1000 μg/mL
Mouse
Primary
Human
ELISA, Immunohistochemistry, Western Blot
1F2B2
Unconjugated
20 μL
23408
IgG2b