Learn More
SIRT5 Mouse anti-Human, Unconjugated, Clone: 1F2B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67257-1-IG-150UL
Additional Details : Poids : 0.00010kg
Description
The SIRT5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SIRT5. This antibody recognizes human antigen. The SIRT5 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SIRT5 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1F2B2 | |
Unconjugated | |
150 μL | |
23408 | |
IgG2b |