Learn More
SLC5A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21628-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Prevents intracellular accumulation of high concentrations of myo-inositol (an osmolyte) that result in impairment of cellular function.Spécification
SLC5A3 | |
Polyclonal | |
Unconjugated | |
SLC5A3 | |
SLC5A3, SMIT, SMIT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114507, 53881, 6526 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P53794, Q9JKZ2 | |
Slc5a3 | |
SLC5A3 Fusion Protein Ag16218 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |