missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLCO4A1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26399-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Mediates the Na(+)-independent transport of organic anions such as the thyroid hormones T3 (triiodo-L-thyronine), T4 (thyroxine) and rT3, and of estrone-3-sulfate and taurocholate.Spécification
SLCO4A1 | |
Polyclonal | |
Unconjugated | |
SLCO4A1 | |
OATP E, OATP RP1, OATP1, OATP4A1, OATPE, OATPRP1, POAT, SLC21A12, SLCO4A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28231 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96BD0 | |
SLCO4A1 | |
SLCO4A1 Fusion Protein Ag23905 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |