missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX22 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13062-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a sorting nexin that is found in the cytoplasm, where it interacts with membrane-bound phosphatidylinositol 3-phosphate. The encoded protein may play a role in intracellular trafficking. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.Spécification
SNX22 | |
Polyclonal | |
Unconjugated | |
SNX22 | |
SNX22, sorting nexin 22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
382083, 79856 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96L94 | |
Snx22 | |
SNX22 Fusion Protein Ag3751 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |