missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14010-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Transcriptional activator. Binds specifically to the DNA sequence 5′-AACAAT-3′. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.Spécification
SOX6 | |
Polyclonal | |
Unconjugated | |
SOX6 | |
HSSOX6, SOX6, SOXD, Transcription factor SOX 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20679, 293165, 55553 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35712, P40645 | |
SOX6 | |
SOX6 Fusion Protein Ag5111 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |