missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTLC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 51012-2-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a long chain base subunit of serine palmitoyltransferase. Serine palmitoyltransferase, which consists of two different subunits, is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine. Mutations in this gene were identified in patients with hereditary sensory neuropathy type I. Alternatively spliced variants encoding different isoforms have been identified.Spécification
SPTLC2 | |
Polyclonal | |
Unconjugated | |
SPTLC2 | |
KIAA0526, LCB 2, LCB2, LCB2a, Serine palmitoyltransferase 2, SPT 2, SPT2, SPTLC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20773, 9517 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15270, P97363 | |
SPTLC2 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |