missing translation for 'onlineSavingsMsg'
Learn More

STK38L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 12697-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16890554

  • 129.00€ / 20µL
Expédition estimée: 13-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to STK38L.

STK38L contains 12 protein kinase catalytic subdomains and a potential bipartite nuclear localization signal. Northern blot analysis revealed expression of a 3.9-kb transcript in all tissues tested, with the possible exception of adult brain. Highest expression was in peripheral blood leukocytes. Immunoblot and immunofluorescence microscopy demonstrated predomitly nuclear expression of a 55-kD protein.
Spécification

Spécification

STK38L
Polyclonal
Unconjugated
STK38L
KIAA0965, NDR2, NDR2 protein kinase, Nuclear Dbf2 related kinase 2, STK38L
Rabbit
Antigen Affinity Chromatography
RUO
23012, 232533, 691337
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7TSE6, Q9Y2H1
Stk38l
STK38L Fusion Protein Ag3413
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions