missing translation for 'onlineSavingsMsg'
Learn More

SUB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10948-2-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837673

  • 399.00€ / 150µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody recognize 14-16 kDa (P14 or P15), 20 kDa (phospho form) and 30 kDa (Dimer form).

PC4 (activated RNA polymerase II transcription cofactor 4) is a transcriptional coactivator, possessing the ability to suppress promoter-driven as well as nonspecific transcription via its DNA binding activity. The repressive activity of PC4 on promoter-driven transcription is alleviated by transcription factor TFIIH. TFIIH protects promoters from PC4-mediated repression by relieving the topological constraint imposed by PC4 through the ERCC3 helicase activity.
Spécification

Spécification

SUB1
Polyclonal
Unconjugated
SUB1
p14, PC4, Positive cofactor 4, RPO2TC1, SUB1, SUB1 homolog, SUB1 homolog (S. cerevisiae)
Rabbit
Antigen Affinity Chromatography
RUO
10923, 192269, 20024
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P11031, P53999, Q63396
Sub1
SUB1 Fusion Protein Ag1390
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions