Learn More
Synaptoporin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14143-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16832654
  • 140.00€ / 20µL
Expédition estimée: 12-04-2024
Description
Synaptophysin 2, also known as synaptoporin, is a component of the synaptic vesicle membrane and thought to play an important role in synaptic vesicle trafficking (Dai et al., 2003). It is highly homologous to synaptophysin 1, yet differs in expression. Whereas synaptophysin 1 is ubiquitously expressed, synaptophysin 2 is concentrated in the mossy fiber synapses of the hippocampus (Grabs et al., 1994; Singec et al., 2002).
Spécification
Spécification
Synaptoporin
Polyclonal
Unconjugated
SYNPR
SPO, synaptoporin, SYNPR
Rabbit
Antigen Affinity Chromatography
RUO
132204, 72003
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BGN8, Q8TBG9
Synpr
Synaptoporin Fusion Protein Ag5307
20 μL
Primary
Human, Mouse