missing translation for 'onlineSavingsMsg'
Learn More

Syntaxin 16 Mouse anti-Human, Clone: 4A9D8, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66775-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16818965

  • 399.00€ / 150µL
Expédition estimée: 03-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a protein that is a member of the syntaxin or t-SNARE (target-SNAP receptor) family. These proteins are found on cell membranes and serve as the targets for V-SNARES (vesicle-SNAP receptors) permitting specific synaptic vesicle docking and fusion. A microdeletion in the region of chromosome 20 where this gene is located has been associated with pseudohypoparathyroidism type Ib. Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

Syntaxin 16
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14662
STX16
Syntaxin 16 Fusion Protein Ag27926
150 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
4A9D8
Unconjugated
STX16
hsyn16, STX16, SYN16, syntaxin 16
Mouse
Protein G
RUO
8675
-20°C
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions