missing translation for 'onlineSavingsMsg'
Learn More

Syntaxin 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14988-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16833944

  • 325.38€ / 150µL
Expédition estimée: 10-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

STX4 is a plasma membrane t-SNARE that mediates docking of transport vesicles.STX4 is necessary for the translocation of SLC2A4 from intracellular vesicles to the plasma membrane. Together with STXB3 and VAMP2, STX4 may also play a role in docking/fusion of intracellular GLUT4-containing vesicles with the cell surface in adipocytes. STX4 may also play a role in docking of synaptic vesicles at presynaptic active zones.
Spécification

Spécification

Syntaxin 4
Polyclonal
Unconjugated
STX4
p35 2, STX4, STX4A, syntaxin 4
Rabbit
Antigen Affinity Chromatography
RUO
20909, 6810, 81803
-20°C
Liquid
Western Blot, Immunoprecipitation
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P70452, Q08850, Q12846
STX4
Syntaxin 4 Fusion Protein Ag6973
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions