Learn More
TARS Mouse, anti-Human, Unconjugated, Clone: 1A11F10, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67828-1-IG-150UL
Informations supplémentaires : Poids : 0.00020kg
 Afficher plus de versions de ce produit
Code produit. 17281476
  • 349.00€ / 150µL
Expédition estimée: 09-10-2023
pour voir le stock disponible 
Description
Description
TARS (Threonyl-tRNA synthetase), also known as threonine-tRNA ligase or ThrRS, belongs to the aminoacyl-tRNA synthetase family. Aminoacyl-tRNA synthetase (aaRS) catalyzes the specific esterification of an amino acid with the 3′-terminal hydroxyl group of its cognate tRNA. Specificity of aminoacylation catalyzed by aaRSs ensures the accurate translation of the genetic code in the first step of protein synthesis (PMID: 26324710). TARS has some isoforms with the molecular mass of 83, 87 kDa and other MW.
Spécification
Spécification
TARS
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat
Western Blot, ELISA
1A11F10
Unconjugated
150 μL
6897