Learn More
TGFBR2 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 27212-1-AP-150UL
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17261386
  • 325.38€ / 150µL
Expédition estimée: 10-10-2022
Description
TGF beta Receptor II (TGFBR2) is a member of the transforming growth factor-β (TGF-β) superfamily, which are critical regulators of cell proliferation and differentiation, developmental patterning and morphogenesis, and disease pathogenesis (PMID: 10974075). TGFBR2 result in both SMAD4-dependent constraint of proliferation and SMAD4-independent activation of apoptosis (PMID: 29396446). It has some isoforms with the molecular weight of 70-80 kDa and ∼90 kDa after glycosylated.
Spécification
Spécification
TGFBR2
Polyclonal
Unconjugated
150 μL
7048
Liquid
ELISA, Immunohistochemistry, Western Blot
200 μg/mL