missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THYN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15867-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
THYN1, also named as THY28, is a 28-32 kDa protein which located mainly in the nucleus. It appears to correlate with induction of apoptosis.Spécification
THYN1 | |
Polyclonal | |
Unconjugated | |
THYN1 | |
HSPC144, MDS012, My0054, MY105, THY28, THY28KD, thymocyte nuclear protein 1, Thymocyte protein Thy28, THYN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29087, 300470, 77862 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P3E0, Q91YJ3, Q9P016 | |
THYN1 | |
THYN1 Fusion Protein Ag8481 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |