Learn More
TIAM1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 27694-1-AP-150UL
Informations supplémentaires : Poids : 0.00020kg
 Afficher plus de versions de ce produit
Code produit. 17281386
  • 349.00€ / 150µL
Expédition estimée: 17-10-2023
pour voir le stock disponible 
Description
Description
The TIAM1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TIAM1. This antibody recognizes human antigen. The TIAM1 antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
Spécification
TIAM1
Polyclonal
Unconjugated
150 μL
7074
Liquid
Western Blot, ELISA
1200 μg/mL