missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20718-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The antibody is specific to TMC1.
This gene is considered a member of a gene family predicted to encode transmembrane proteins. The specific function of this gene is unknown; however, it is known to be required for normal function of cochlear hair cells. Mutations in this gene have been associated with progressive postlingual hearing loss and profound prelingual deafness.Spécification
TMC1 | |
Polyclonal | |
Unconjugated | |
TMC1 | |
DFNA36, DFNB11, DFNB7, TMC1, transmembrane channel like 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117531, 13409, 361739 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R4P5, Q8TDI8 | |
Tmc1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |