missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12196-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is involved in the import of precursor proteins into mitochondria. The encoded protein has a chaperone-like activity, binding the mature portion of unfolded proteins and aiding their import into mitochondria. This protein, which is found in the cytoplasm and sometimes associated with the outer mitochondrial membrane, has a weak ATPase activity and contains 6 TPR repeats.Spécification
TOMM34 | |
Polyclonal | |
Unconjugated | |
TOMM34 | |
hTom34, HTOM34P, TOM34, TOMM34, URCC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10953, 311621, 67145 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15785, Q3KRD5, Q9CYG7 | |
Tomm34 | |
TOMM34 Fusion Protein Ag2834 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |