missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAcP (Tartrate-Resistant Acid Phosphatase) (Hairy Cell Leukemia Marker) Mouse, Clone: rACP5/1070, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 54-MSM3-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
TRAcP (Tartrate-Resistant Acid Phosphatase) (Hairy Cell Leukemia Marker) | |
Cytoplasmic (Lysosomes) | |
rACP5/1070 | |
Unconjugated | |
Mouse | |
35kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
54 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human ACP5 protein | |
HepG2, 293T, K562 or RPMI-8226 Cells. Spleen from Hairy Cell Leukemia (HCL) patient. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |