missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transcription Termination Factor Mouse anti-Human, Mouse, Clone: 7B3H2, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60235-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The antibody is specific to TTF1.
TTF1 is a transcription factor that binds and activates the promoter of thyroid specific genes such as thyroglobulin, thyroperoxidase, and thyrotropin receptor. Crucial in the maintece of the thyroid differentiation phenotype. May play a role in lung development and surfactant homeostasis.Spécification
Transcription Termination Factor | |
Monoclonal | |
2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15361, Q62187 | |
TTF1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Western Blot | |
7B3H2 | |
Unconjugated | |
TTF1 | |
TTF 1, TTF I, TTF1 | |
Mouse | |
Protein A | |
RUO | |
22130, 7270 | |
-20°C | |
Liquid |